Trimethylsilyl cyanide, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7677-24-9
:
Formula:C₄H₉NSi
MDL Number:MFCD00001765
:99.21 g/mol
Boiling Pt:114 °C (1013 hPa)
Melting Pt:11 °C
Density:0.79 g/cm³ (20 °C)
Flash Pt:1 °C
UN:3383
ADR:6.1,I
:M-Clarity™ quality level = MQ200
EINECS:231-657-3
Frequently Bought Together