1-Adamantyl isothiocyanate, Sigma-Aldrich®
Catalog # SIAL143367-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
Boiling Pt:270 °C (1013 hPa)
Melting Pt:174…175 °C
:193.31 g/mol
EINECS:224-564-4
CAS Number:4411-26-1
Formula:C₁₁H₁₅NS
MDL Number:MFCD00074736