3,3',5,5'-Tetramethylbenzidine (TMB)
Glass bottle for solids, 5 g
Catalog # MOLEM88604857
Marketsource
Supplier: Molekula
3,3',5,5'-Tetramethylbenzidine (TMB)
Catalog # MOLEM88604857
Supplier: Molekula
Marketsource
CAS Number: 54827-17-7
Jump to:
Product Details & Documents
Synonyms: TMB, 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine
- CAS Number:54827-17-7
- Formula:C₁₆H₂₀N₂
- MDL Number:MFCD00007748
- Molecular Weight:240.35 g/mol
- Boiling Pt:402 °C (1013 hPa)
- Melting Pt:166…170 °C
- Storage Temperature:Ambient
- EINECS:259-364-6
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



