Tetrakis(triphenylphosphine)palladium(0)
Catalog # MOLE10389753-1G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:1155.58 g/mol
Melting Pt:103…107 °C
Density:1 g/cm³ (20 °C)
EINECS:238-086-9
CAS Number:14221-01-3
Formula:C₇₂H₆₀P₄Pd
MDL Number:MFCD00010012