Order Entry
Poland
ContactUsLinkComponent
5-(2-Bromophenyl)-1H-tetrazole
5-(2-Bromophenyl)-1H-tetrazole
Catalog # MOLEM85598531
Supplier:  Molekula
CAS Number:  
5-(2-Bromophenyl)-1H-tetrazole
Catalog # MOLEM85598531
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :225.05 g/mol
Melting Pt:183…184 °C
CAS Number:73096-42-1
Formula:C₇H₅BrN₄
MDL Number:MFCD00151799