Thiophene-2-carboxylic hydrazide
Catalog # MOLEM73362117
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:142.18 g/mol
Melting Pt:142…143 °C
EINECS:219-107-0
CAS Number:2361-27-5
Formula:C₅H₆N₂OS
MDL Number:MFCD00005435