Order Entry
Poland
ContactUsLinkComponent
Thiophene-2-carbaldehyde
Irritant
Thiophene-2-carbaldehyde
Catalog # MOLEM47706578
Supplier:  Molekula
CAS Number:  
Irritant
Thiophene-2-carbaldehyde
Catalog # MOLEM47706578
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :112.15 g/mol
Boiling Pt:198 °C (1013 hPa)
Melting Pt:? 10 °C
Density:1,2264 g/cm³ (20 °C)
Flash Pt:78 °C
EINECS:202-629-8
 :GHS07
CAS Number:98-03-3
Formula:C₅H₄OS
MDL Number:MFCD00005429