Order Entry
Poland
ContactUsLinkComponent
Sodium bisulphite 39% in aqueous solution for synthesis, Sigma-Aldrich®
Sodium bisulphite 39% in aqueous solution for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Sodium bisulphite 39% in aqueous solution for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    -44 °C
  • Boiling Point:
    146 °C
  • Vapor Pressure:
    40 hPa (20 °C)
  • Density:
    1.34 g/cm3 (20 °C)
  • pH:
    4.5 (H2O, 20 °C)
  • Hazard Class:
    8
  • Hazard Class Description:
    Corrosive material

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together