Order Entry
Poland
ContactUsLinkComponent
 
 
2-Aminobenzothiazole, Sigma-Aldrich®
 :  Merck
 
 
2-Aminobenzothiazole, Sigma-Aldrich®
 :  Merck

 

  • Solubility:
    (20 °C) insoluble
  • Melting Point:
    127 - 130 °C
  • Molar Mass:
    150.2 g/mol
  • Synonyms:
    2-Benzothiazolylamine
  • Hill Formula:
    C7H6N2S

 

Frequently Bought Together