Order Entry
Poland
ContactUsLinkComponent
5-(3-Chlorophenyl)-1H-tetrazole ≥98%
Irritant
5-(3-Chlorophenyl)-1H-tetrazole ≥98%
Catalog # L10699.03
CAS Number:  
Irritant
5-(3-Chlorophenyl)-1H-tetrazole ≥98%
Catalog # L10699.03
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

CAS Number:41421-28-7
 :
Formula:C₇H₅ClN₄
 :180.6 g/mol
Melting Pt:139…140 °C
 :GHS07
Storage Temperature:Ambient
MDL Number:MFCD00040951