Złóż zamówienie
Poland
Orders LinkContactUsLinkComponent
 

3,3-Dimetylocykloheksanon ≥97%

Glass bottle for solids, 5 g
Catalog # H28000.06
Flammable
3,3-Dimetylocykloheksanon ≥97%
Catalog # H28000.06
CAS Number:  2979-19-3
3,3-Dimetylocykloheksanon ≥97%
3,3-Dimetylocykloheksanon ≥97%

Glass bottle for solids, 5 g

CAS Number:  2979-19-3
Price
886 zł
Log in for availability
1 * 5 g
Product Details & Documents

Purity: 97% CAS No.: 2979-19-3 Molecular Formula: C8H14O Formula Weight: 126.20 Flash Point: 59°(138°F) Refractive Index: 1.4470 Boiling Point: 175° UN No.: 1224 MDL No.: MFCD00040178

  • CAS Number:2979-19-3
  • Formula:C₈H₁₄O
  • MDL Number:MFCD00040178
  • Molecular Weight:126.2 g/mol
  • Boiling Pt:175 °C
  • Melting Pt:6 °C
  • Density:0,91
  • Flash Pt:59 °C (138 °F)
  • Storage Temperature:Temperatura otoczenia
  • UN:1224
  • ADR:3,III
  •  :Density: 0.909 g/cm³ (25 °C)
Specifications

Specifications

  • Rodzaj opakowania:
    Glass bottle for solids
  • Ilość:
    5 g

Specifications