Order Entry
Poland
ContactUsLinkComponent
Dicyclopentadiene ≥95% stabilised
Flammable
Irritant
Environment
Dicyclopentadiene ≥95% stabilised
Catalog # 32546.30
CAS Number:  
Flammable
Irritant
Environment
Dicyclopentadiene ≥95% stabilised
Catalog # 32546.30
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g
  • Seq:
    3545

Specifications

About this item

 :
 :132.21 g/mol
Merck Index:14,2739
Boiling Pt:171…173 °C
Melting Pt:12…34 °C
Density:0.982
Flash Pt:26 °C (78 °F)
Storage Temperature:Ambient
EINECS:201-052-9
UN:2048
ADR:3,III
 :Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
 :GHS02
 :GHS07
 :GHS09
CAS Number:77-73-6
Formula:C₁₀H₁₂
MDL Number:MFCD00078246