


Trimethylsilyl cyanide 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7677-24-9
:
Formula:C₄H₉NSi
MDL Number:MFCD00001765
:99.21 g/mol
Boiling Pt:114 °C (1013 hPa)
Melting Pt:11 °C
Density:0.744 g/cm³
Flash Pt:1 °C
Storage Temperature:Refrigerator
EINECS:231-657-3
UN:3384
ADR:6.1,I
:GHS06
:GHS09
:GHS02
Specification Test Results
- Appearance:Clear colorless to yellow-brown Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3900 to 1.3930 (20°C, 589 nm)
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together