cis-4-Hydroxy-D(+)-proline, Sigma-Aldrich®
Catalog # SIALH5877-10G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:131.13 g/mol
:M-Clarity™ quality level = MQ100
EINECS:219-963-5
CAS Number:2584-71-6
Formula:C₅H₉NO₃
MDL Number:MFCD00005252