Order Entry
Poland
ContactUsLinkComponent
Sodium-L(+)-glutamate monohydrate 98.5-101.5% (dried basis) FCC, J.T.Baker®
Sodium-L(+)-glutamate monohydrate 98.5-101.5% (dried basis) FCC, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Sodium-L(+)-glutamate monohydrate 98.5-101.5% (dried basis) FCC, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Food GMP Manufactured Product:
    Passes Test
  • Meets F.C.C. Requirements:
    98.5 - 101.5%
  • CAUTION: For Manufacturing, processing or repackaging:
    ≤0.2%
  • Bulk Food Chemical:
    ≤5 mg/kg
  • FCC - Identification:
    Passes Test
  • FCC - Assay (dried basis):
    98.5 - 101.5 %
  • FCC - Chloride (Cl):
    ≤0.2 %
  • FCC - Lead (Pb):
    ≤5 mg/kg
  • FCC - Clarity and Color of Solution:
    Passes Test
  • FCC - Loss on Drying:
    ≤0.5 %
  • FCC - Specific Rotation [Å]²ºD (+):
    24.5 - 25.5 Degree

Specification Test Results

Related Information

Frequently Bought Together