
1-(4-Aminophenyl)-3,3,3-trifluoropropan-1-one 95%
Catalog # APOSPC50739-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:203.161 g/mol
:GHS07
CAS Number:7119-16-6
Formula:C₉H₈F₃NO
MDL Number:MFCD19673474