
3'-(Trifluoromethyl)-2,2,2,4'-tetrafluoroacetophenone 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1335013-55-2
:
Formula:C₉H₃F₇O
MDL Number:MFCD22688446
:260.11 g/mol
:GHS07
Frequently Bought Together
Water, AnalaR NORMAPUR® ISO 3696 Grade 3 analytical reagent
Supplier: VWR Chemicals
Glycerine 85% Ph. Eur.
Supplier: VWR Chemicals
Ethanol absolute ≥99.5%, EMPLURA®, Supelco®
Supplier: Merck
Toluene ≥99.90%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Dichloromethane ≥99.9% stabilised, PESTINORM® Supra Trace for organic trace analysis
Supplier: VWR Chemicals
Ethyl acetate ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
HYDROCARBONS C6-11 HYDROTREATED 1 * 5 L
Supplier: CHEMPUR
Ethanol 96% ≥96% (v/v), TechniSolv®, pure
Supplier: VWR Chemicals
Ethanol absolute ≥99.5%, TechniSolv®, pure
Supplier: VWR Chemicals