
[1,3-Bis(2,6-Diisopropylphenyl)imidazol-2-ylidene](3-chloropyridyl)palladium(II) dichloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:905459-27-0
:
Formula:C₃₂H₄₀Cl₃N₃Pd
MDL Number:MFCD08457656
Melting Pt:230 °C
:679.46 g/mol
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together