Order Entry
Poland
ContactUsLinkComponent
Diethylene glycol monododecyl ether 95%
Irritant
Diethylene glycol monododecyl ether 95%
Catalog # APOSOR938002-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Diethylene glycol monododecyl ether 95%
Catalog # APOSOR938002-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :274.45 g/mol
Melting Pt:16 °C
 :GHS07
CAS Number:3055-93-4
Formula:C₁₆H₃₄O₃
MDL Number:MFCD00042658