2-Ethylhexyl-4-hydroxybenzoate 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:5153-25-3
:
Formula:C₁₅H₂₂O₃
MDL Number:MFCD00016472
:250.34 g/mol
Boiling Pt:362.6/760 mm °C
Melting Pt:118…120 °C
Density:1.035
Flash Pt:144.6 °C
Frequently Bought Together