Order Entry
Poland
ContactUsLinkComponent
2-[3-(Methylsulphonyl)phenyl]ethylamine 95%
Irritant
2-[3-(Methylsulphonyl)phenyl]ethylamine 95%
Catalog # APOSOR950154-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-[3-(Methylsulphonyl)phenyl]ethylamine 95%
Catalog # APOSOR950154-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :199.27 g/mol
 :GHS07
CAS Number:933696-63-0
Formula:C₉H₁₃NO₂S
MDL Number:MFCD13190314