Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Buffers
Cloning Buffers
Buffer, sodium acetate buffer solution pH 5,2 (3 mol/l), Millipore®
Buffer, sodium acetate buffer solution pH 5,2 (3 mol/l), Millipore®
Buffer, sodium acetate buffer solution pH 5,2 (3 mol/l), Millipore®
567422-100
:
Merck Millipore (Calbiochem)
:
Buffer, sodium acetate buffer solution pH 5,2 (3 mol/l), Millipore®
567422-100
:
Merck Millipore (Calbiochem)
:
Pack type:
Glass bottle
Pk:
100 mL
Description:
Sodium acetate buffer solution pH 5,2 (3 mol/l)