
N-(3-Triethylammoniumpropyl)-4-(4-(dibutylamino)styryl)pyridinium dibromide (MM 1-43) ≥95%
Catalog # AATB21483
Supplier: AAT Bioquest
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 mg
- Seq:2070
Specifications
About this item
:
:611.54 g/mol
ADR:5,II
:GHS07
CAS Number:149838-22-2
Formula:C₃₀H₄₉Br₂N₃
MDL Number:MFCD00467815