Water, colourless
Bottle, 1 L
Catalog # SIALTMS-011-A
Supplier: Merck
Jump to:
Product Details & Documents
Synonyms: Hydrogen oxide
- CAS Number:7732-18-5
- Formula:H₂O
- MDL Number:MFCD00011332
- Molecular Weight:18.02 g/mol
- Boiling Pt:100,0 °C (1.013 hPa)
- Melting Pt:0,0 °C
- Density:1,000 g/cm3
Specifications
Specifications
- Pack type:Bottle
- Pk:1 L
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



