Order Entry
Poland
ContactUsLinkComponent
Cyanine 5.5 maleimide ≥90% [equivalent to Cy5.5® maleimide]
Cyanine 5.5 maleimide ≥90% [equivalent to Cy5.5® maleimide]
Catalog # AATB175
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Cyanine 5.5 maleimide ≥90% [equivalent to Cy5.5® maleimide]
Catalog # AATB175
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :1153.2 g/mol