Order Entry
Poland
Orders LinkContactUsLinkComponent
Pro-AMC

Pro-AMC

Plastic bottle, 5 mg
Catalog # AATB13457
Irritant
Supplier:   AAT Bioquest
Pro-AMC
Catalog # AATB13457
Supplier:  AAT Bioquest
CAS Number:  
Pro-AMC
Pro-AMC

Plastic bottle, 5 mg

CAS Number:  
Price
664 zł
Log in for availability
1 * 5 mg
Product Details & Documents

Pro-AMC is a sensitive fluorogenic protease substrate.

  • Protease substrate
  • Bright signal
  • Multiple applications
  • ADR:5,II
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.