Order Entry
Poland
Orders LinkContactUsLinkComponent
Z-DEVD-ProRed™ 620

Z-DEVD-ProRed™ 620

Plastic bottle, 1 mg
Catalog # AATB13433
Irritant
Supplier:   AAT Bioquest
Z-DEVD-ProRed™ 620
Catalog # AATB13433
Supplier:  AAT Bioquest
CAS Number:  
Z-DEVD-ProRed™ 620
Z-DEVD-ProRed™ 620

Plastic bottle, 1 mg

CAS Number:  
Price
2 000 zł
Log in for availability
1 * 1 mg
Product Details & Documents

ProRed™-derived protease substrates are colorless and non-fluorescent.

  • Caspase detection
  • Bright signal
  • ADR:5,II
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.