Order Entry
Poland
ContactUsLinkComponent
Recombinant Mouse THPO (from HEK293 cells), His-tag
Recombinant Mouse THPO (from HEK293 cells), His-tag
Catalog # ABNOP9291.10UG
Supplier:  Abnova
CAS Number:  
Recombinant Mouse THPO (from HEK293 cells), His-tag
Catalog # ABNOP9291.10UG
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    10 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Mouse
  • Environmentally Preferable:
  • Tag sequence:
    His-tag
  • Protein/peptide name:
    THPO

Specifications

About this item