Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
Recombinant Sheep PRL (from E. coli), His-tag
Recombinant Sheep PRL (from E. coli), His-tag
Recombinant Sheep PRL (from E. coli), His-tag
ABNOP9053.50UG
:
Abnova
:
Recombinant Sheep PRL (from E. coli), His-tag
ABNOP9053.50UG
:
Abnova
:
Pk:
50 µG
Protein/peptide type:
Recombinant
Source:
E. coli
Species:
Sheep
Tag sequence:
His-tag
Gene ID:
P01240
Protein/peptide name:
PRL