Recombinant Human CD5 (from E. coli), His-tag
Catalog # ABNOP9643.20UG
Supplier: Abnova
CAS Number:
Specifications
- Pk:20 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Environmentally Preferable:
- Tag sequence:His-tag
- Gene ID:P06127
- Protein/peptide name:CD5
- Sequence:25a.a.-372a.a.