Recombinant Human CD207 (from E. coli), His-tag
Catalog # ABNOP9351.20UG
Supplier: Abnova
CAS Number:
Specifications
- Pk:20 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Environmentally Preferable:
- Tag sequence:His-tag
- Protein/peptide name:CD207