Order Entry
Poland
ContactUsLinkComponent
Recombinant Mouse CCL3 (from E. coli), His-tag
Recombinant Mouse CCL3 (from E. coli), His-tag
Catalog # ABNOP9537.10UG
Supplier:  Abnova
CAS Number:  
Recombinant Mouse CCL3 (from E. coli), His-tag
Catalog # ABNOP9537.10UG
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    10 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Environmentally Preferable:
  • Tag sequence:
    His-tag
  • Gene ID:
    P10855
  • Protein/peptide name:
    CCL3
  • Sequence:
    24a.a.-92a.a.

Specifications

About this item