Order Entry
Poland
ContactUsLinkComponent
Recombinant Human TNF (from E. coli), His-tag
Recombinant Human TNF (from E. coli), His-tag
Catalog # ABNOP9222.50UG
Supplier:  Abnova
CAS Number:  
Recombinant Human TNF (from E. coli), His-tag
Catalog # ABNOP9222.50UG
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    His-tag
  • Protein/peptide name:
    TNF

Specifications

About this item