Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Wprowadź zamówienie
Status zamówienia
Twoje konto
Koszyk
Poland
Wprowadź zamówienie
Status zamówienia
Twoje konto
Koszyk
ContactUsLinkComponent
Home
Proteins and Peptides
Recombinant Human TNFRSF1B (from CHO cells)
Recombinant Human TNFRSF1B (from CHO cells)
Recombinant Human TNFRSF1B (from CHO cells)
Catalog # ABNOP9248.50UG
Supplier:
Abnova
CAS Number:
Expand image
Recombinant Human TNFRSF1B (from CHO cells)
Catalog # ABNOP9248.50UG
Supplier:
Abnova
CAS Number:
Specifications
Pk:
50 µG
Protein/peptide type:
Recombinant
Source:
CHO cells
Species:
Human
Protein/peptide name:
TNFRSF1B
Specifications
About this item
Product Details
Product Details