Order Entry
Poland
ContactUsLinkComponent
Recombinant Human TNFRSF10A (from E. coli), Fc tag
Recombinant Human TNFRSF10A (from E. coli), Fc tag
  ABNOP9261.50UG
 :  Abnova
 :  
Recombinant Human TNFRSF10A (from E. coli), Fc tag
  ABNOP9261.50UG
 :  Abnova
 :  

 

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    Fc tag
  • Protein/peptide name:
    TNFRSF10A