Order Entry
Poland
Orders LinkContactUsLinkComponent
Recombinant Human SAA4 (from E. coli), His-tag

Recombinant Human SAA4 (from E. coli), His-tag

10 µG
Catalog # ABNOP9177.10UG
Supplier:   Abnova
Recombinant Human SAA4 (from E. coli), His-tag
Catalog # ABNOP9177.10UG
Supplier:  Abnova
CAS Number:  
Recombinant Human SAA4 (from E. coli), His-tag

10 µG

CAS Number:  
Price
1 120 zł
Log in for availability
1 * 10 µG
Product Details & Documents
Specifications

Specifications

  • Pk:
    10 µG
  • Cat. No.:
    ABNOP9177.10UG
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    SAA4
  • Tag sequence:
    His-tag
  • Species:
    Human
  • Source:
    E. coli

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.