Recombinant Mouse ARSA (from Baculovirus), His-tag
ABNOP8053.50UG
: Abnova
:
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:Baculovirus
- Species:Mouse
- Tag sequence:His-tag
- Gene ID:P50428
- Protein/peptide name:ARSA
- Sequence:18a.a.-506a.a.