Order Entry
Poland
ContactUsLinkComponent
Recombinant Mouse CD200R1 (from Baculovirus), hIgG-His-tag
Recombinant Mouse CD200R1 (from Baculovirus), hIgG-His-tag
  ABNOP7991.100UG
 :  Abnova
 :  
Recombinant Mouse CD200R1 (from Baculovirus), hIgG-His-tag
  ABNOP7991.100UG
 :  Abnova
 :  

 

  • Pk:
    100 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Baculovirus
  • Species:
    Mouse
  • Environmentally Preferable:
  • Tag sequence:
    hIgG-His-tag
  • Gene ID:
    Q9ES57
  • Protein/peptide name:
    CD200R1
  • Sequence:
    261a.a.-238a.a.