Order Entry
Poland
ContactUsLinkComponent
 
Recombinant Human ATP1B1 (from Baculovirus), His-tag
  ABNOP8003.250UG
 :  Abnova
 :  
Recombinant Human ATP1B1 (from Baculovirus), His-tag
  ABNOP8003.250UG
 :  Abnova
 :  

 

  • Pk:
    250 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Baculovirus
  • Species:
    Human
  • Environmentally Preferable:
  • Tag sequence:
    His-tag
  • Gene ID:
    P05026
  • Protein/peptide name:
    ATP1B1
  • Sequence:
    63a.a.-303a.a.