Order Entry
Poland
ContactUsLinkComponent
Recombinant Human GPT (from E. coli), His-tag
Recombinant Human GPT (from E. coli), His-tag
  ABNOP7949.50UG
 :  Abnova
 :  
Recombinant Human GPT (from E. coli), His-tag
  ABNOP7949.50UG
 :  Abnova
 :  

 

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    His-tag
  • Gene ID:
    P24298
  • Protein/peptide name:
    GPT
  • Sequence:
    1a.a.-496a.a.