Recombinant Mouse PDCD1 (from HEK293 cells), hFc tag
ABNOP7548.1MG
: Abnova
:
- Pk:1 mg
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Environmentally Preferable:
- Tag sequence:hFc tag
- Gene ID:Q02242
- Protein/peptide name:PDCD1
- Sequence:25a.a.-167a.a.