Order Entry
Poland
ContactUsLinkComponent
Recombinant Human CD24 (from HEK293 cells), hFc tag
Recombinant Human CD24 (from HEK293 cells), hFc tag
Catalog # ABNOP7618.50UG
Supplier:  Abnova
CAS Number:  
Recombinant Human CD24 (from HEK293 cells), hFc tag
Catalog # ABNOP7618.50UG
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Tag sequence:
    hFc tag
  • Gene ID:
    P25063
  • Protein/peptide name:
    CD24
  • Sequence:
    27a.a.-59a.a.

Specifications

About this item