Recombinant Human CSF2 (from P. pastoris)
10 µG
Catalog # ABNOP7136.10UG
Supplier: Abnova
Jump to:
Product Details & Documents
Specifications
Specifications
- Pk:10 µG
- Cat. No.:ABNOP7136.10UG
- Protein/peptide type:Recombinant
- Protein/peptide name:CSF2
- Gene ID:P04141
- Sequence:18a.a.-144a.a.
- Species:Human
- Source:P. pastoris



