Recombinant Human IL10 (from CHO cells)
Catalog # ABNOP7102.10UG
Supplier: Abnova
CAS Number:
Specifications
- Pk:10 µG
- Protein/peptide type:Recombinant
- Source:CHO cells
- Species:Human
- Environmentally Preferable:
- Gene ID:P22301
- Protein/peptide name:IL10
- Sequence:19a.a.-178a.a.