Anti-TP53 Rabbit Recombinant Antibody [clone: P53S15-1C11] (APC)
Catalog # ABNORAB02895.100
Supplier: Abnova
Specifications
- Antigen symbol:TP53
- Clonality:Recombinant
- Clone:P53S15-1C11
- Conjugation:APC (Allophycocyanin)
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human TP53.
- Pk:100 AU