Anti-INPPL1 Rabbit Recombinant Antibody [clone: SHIP2Y1135-1D2]
ABNORAB02816.200UL
: Abnova
- Antigen symbol:INPPL1
- Clonality:Recombinant
- Clone:SHIP2Y1135-1D2
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human INPPL1.
- Pk:200 µl