Anti-STAT1 Rabbit Recombinant Antibody [clone: STAT1S727-C6]
Catalog # ABNORAB02829.200UL
Supplier: Abnova
Specifications
- Antigen symbol:STAT1
- Clonality:Recombinant
- Clone:STAT1S727-C6
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human STAT1.
- Pk:200 µl