Anti-ANXA1 Rabbit Recombinant Antibody [clone: SAIC-14C-10F12]
Catalog # ABNORAB03665.200UG
Supplier: Abnova
Specifications
- Antigen symbol:ANXA1
- Clonality:Recombinant
- Clone:SAIC-14C-10F12
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human ANXA1.
- Pk:200 µG