Anti-EGFR Rabbit Recombinant Antibody [clone: EGFRY1068-E5]
Catalog # ABNORAB02794.200UL
Supplier: Abnova
Specifications
- Antigen symbol:EGFR
- Clonality:Recombinant
- Clone:EGFRY1068-E5
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human EGFR.
- Pk:200 µl