Anti-LCP2 Rabbit Recombinant Antibody [clone: SLP76Y128-3F8]
Catalog # ABNORAB02802.200UL
Supplier: Abnova
Specifications
- Antigen symbol:LCP2
- Clonality:Recombinant
- Clone:SLP76Y128-3F8
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human LCP2.
- Pk:200 µl